Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sel1l2
Entrez Gene ID
228684
Gene Name
sel-1 suppressor of lin-12-like 2 (C. elegans)
Gene Aliases
Gm118
Chromosome Location
Chr.2: 140229599 - 140389716 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033296.2 NP_001028468.1 9 - 10 985 122
GenBank mRNA AK132650.1 - 9 - 10 986 122
BC150948.1 - 9 - 10 957 122

View Full Product Details