Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Obp2b
Entrez Gene ID
383678
Gene Name
odorant binding protein 2B
Gene Aliases
Lcn14, OTTMUSG00000012631
Chromosome Location
Chr.2: 25714105 - 25740115 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099301.1 NP_001092771.1 2 - 3 272 93

View Full Product Details