Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ubr3
- Entrez Gene ID
-
68795
- Gene Name
- ubiquitin protein ligase E3 component n-recognin 3
- Gene Aliases
-
1110059H15Rik, 4833421P10Rik, A130030D10Rik, AA409735, AA414972, AA422631, AI646861, AU016126, Zfp650, Znf650
- Chromosome Location
-
Chr.2: 69896969 - 70024010 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC172146.1 |
- |
2 - 3 |
425 |
149 |
DQ924536.1 |
- |
2 - 3 |
736 |
149 |
View Full Product Details