Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adam33
Entrez Gene ID
110751
Gene Name
a disintegrin and metallopeptidase domain 33
Gene Aliases
Adaml
Chromosome Location
Chr.2: 131050817 - 131063814 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163529.1 NP_001157001.1 13 - 14 1709 68
NM_033615.2 NP_291093.2 13 - 14 1709 68
GenBank mRNA AB059632.1 - 13 - 14 1709 68
AB059633.1 - 13 - 14 1709 68
AF386072.1 - 9 - 10 1076 68
AF472524.1 - 13 - 14 1441 68
AK048367.1 - 9 - 10 976 68
AY054984.1 - 13 - 14 1409 68
AY382193.1 - 13 - 14 68
AY382194.1 - 13 - 14 68
AY953137.1 - 8 - 9 984 68
BC138139.1 - 13 - 14 1432 68

View Full Product Details