Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adam33
Entrez Gene ID
110751
Gene Name
a disintegrin and metallopeptidase domain 33
Gene Aliases
Adaml
Chromosome Location
Chr.2: 131050817 - 131063814 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163529.1 NP_001157001.1 4 - 5 637 77
NM_033615.2 NP_291093.2 4 - 5 637 77
GenBank mRNA AB059632.1 - 4 - 5 637 77
AB059633.1 - 4 - 5 637 77
AF472524.1 - 4 - 5 369 77
AY054984.1 - 4 - 5 337 77
AY382193.1 - 4 - 5 368 77
AY382194.1 - 4 - 5 368 77
BC138139.1 - 4 - 5 360 77

View Full Product Details