Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prkcg
Entrez Gene ID
18752
Gene Name
protein kinase C, gamma
Gene Aliases
PKCgamma, Pkcc, Prkcc
Chromosome Location
Chr.7: 3303532 - 3331099 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291434.1 NP_001278363.1 12 - 13 1541 65
NM_011102.4 NP_035232.1 13 - 14 1694 65
GenBank mRNA AK144419.1 - 13 - 14 1696 65
BC111807.1 - 12 - 13 1428 65
L28035.1 - 13 - 14 1772 65
X67129.1 - 13 - 14 1503 65

View Full Product Details