Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubr7
Entrez Gene ID
66622
Gene Name
ubiquitin protein ligase E3 component n-recognin 7 (putative)
Gene Aliases
5730410I19Rik, AA589405, AW557761
Chromosome Location
Chr.12: 102757975 - 102777701 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025666.2 NP_079942.1 5 - 6 552 107
GenBank mRNA AK011968.1 - 5 - 6 423 107
AK017542.1 - 5 - 6 508 107
AK088208.1 - 5 - 6 554 107
AK146308.1 - 5 - 6 553 107
AK149989.1 - 5 - 6 507 107
AK155611.1 - 5 - 6 423 107
AK190790.1 - 4 - 5 286 107
BC051678.1 - 2 - 3 80 107
BC058535.1 - 4 - 5 236 107
BC079531.1 - 4 - 5 249 107
BC094323.1 - 5 - 6 559 107
BC094423.1 - 5 - 6 510 107

View Full Product Details