Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lcn11
Entrez Gene ID
227630
Gene Name
lipocalin 11
Gene Aliases
Gm109
Chromosome Location
Chr.2: 25777017 - 25780279 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100455.2 NP_001093925.1 1 - 2 116 70
GenBank mRNA AY301267.1 - 1 - 2 116 70

View Full Product Details