Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mamdc4
Entrez Gene ID
381352
Gene Name
MAM domain containing 4
Gene Aliases
Gm995
Chromosome Location
Chr.2: 25563110 - 25572158 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081199.1 NP_001074668.2 12 - 13 1584 55

View Full Product Details