Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ncs1
Entrez Gene ID
14299
Gene Name
neuronal calcium sensor 1
Gene Aliases
9430075O15Rik, A730032G13Rik, AI836659, Freq, Mfreq, NCS-1
Chromosome Location
Chr.2: 31245828 - 31295989 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019681.3 NP_062655.1 5 - 6 654 77
GenBank mRNA AF020184.1 - 5 - 6 401 77
AK079136.1 - 4 - 5 341 77
BC059825.1 - 5 - 6 654 77

View Full Product Details