Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Serpina3n
- Entrez Gene ID
-
20716
- Gene Name
- serine (or cysteine) peptidase inhibitor, clade A, member 3N
- Gene Aliases
-
Spi2-2, Spi2.2, Spi2/eb.4
- Chromosome Location
-
Chr.12: 104406675 - 104414334 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_009252.2 |
NP_033278.2 |
1 - 2 |
92 |
61 |
View Full Product Details