Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Abhd12
- Entrez Gene ID
-
76192
- Gene Name
- abhydrolase domain containing 12
- Gene Aliases
-
1500011G07Rik, 6330583M11Rik, AI431047, AW547313
- Chromosome Location
-
Chr.2: 150832493 - 150904745 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK018261.1 |
- |
9 - 10 |
980 |
105 |
View Full Product Details