Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tdp1
Entrez Gene ID
104884
Gene Name
tyrosyl-DNA phosphodiesterase 1
Gene Aliases
2810481F14Rik, 4921509N21Rik, AI838772, AW493413, E430034L06Rik, MLZ-501, SCAN1
Chromosome Location
Chr.12: 99884515 - 99955219 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028354.4 NP_082630.2 15 - 16 1806 99
GenBank mRNA AK013427.1 - 14 - 15 1281 99
AK014855.1 - 15 - 16 1786 99
BC019804.1 - 12 - 13 1130 99
BC065162.1 - 14 - 15 1776 99

View Full Product Details