The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005342.2 | NP_001005342.1 | 4 - 5 | 572 | 139 |
GenBank mRNA | AB098743.2 | - | 4 - 5 | 572 | 139 |
AK043577.1 | - | 1 - 2 | 410 | 139 | |
BC119263.1 | - | 3 - 4 | 446 | 139 |