The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033254.3 | NP_001028426.2 | 2 - 3 | 528 | 64 |
GenBank mRNA | AK158913.1 | - | 2 - 3 | 802 | 64 |
AK158944.1 | - | 2 - 3 | 802 | 64 | |
BC059912.1 | - | 2 - 3 | 335 | 64 | |
BC079572.1 | - | 2 - 3 | 528 | 64 | |
BC150754.1 | - | 2 - 3 | 418 | 64 |