Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Clptm1l
Entrez Gene ID
218335
Gene Name
CLPTM1-like
Gene Aliases
C130052I12Rik, Crr9
Chromosome Location
Chr.13: 73604002 - 73620639 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146047.2 NP_666159.2 6 - 7 1038 85
GenBank mRNA AK032295.1 - 6 - 7 1001 85
AK048357.1 - 6 - 7 935 85
AK156203.1 - 6 - 7 929 85
AK165939.1 - 6 - 7 899 85
BC026562.1 - 6 - 7 739 85

View Full Product Details