Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcdh19
Entrez Gene ID
279653
Gene Name
protocadherin 19
Gene Aliases
B530002L05Rik, Gm717, mKIAA1313
Chromosome Location
Chr.X: 133582860 - 133688993 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105245.1 NP_001098715.1 3 - 4 4343 78
NM_001105246.1 NP_001098716.1 2 - 3 4202 78
GenBank mRNA AK122492.1 - 3 - 4 2086 78
BC118529.1 - 3 - 4 2391 78

View Full Product Details