Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr44
Entrez Gene ID
72404
Gene Name
WD repeat domain 44
Gene Aliases
2610034K17Rik, RAB11BP, RPH11
Chromosome Location
Chr.X: 23692926 - 23806026 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK011669.1 - 2 - 3 445 145
AK041211.1 - 4 - 5 421 145
AK088885.1 - 4 - 5 386 145
BC049191.1 - 4 - 5 324 145
BC068151.1 - 3 - 4 520 145

View Full Product Details