Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr44
Entrez Gene ID
72404
Gene Name
WD repeat domain 44
Gene Aliases
2610034K17Rik, RAB11BP, RPH11
Chromosome Location
Chr.X: 23692926 - 23806026 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC049191.1 - 6 - 7 1115 77
BC068151.1 - 5 - 6 1311 77

View Full Product Details