Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr13
Entrez Gene ID
73447
Gene Name
WD repeat domain 13
Gene Aliases
1700060B08Rik, 5730411P10Rik, DXHXS7467e, W51679, mMg21
Chromosome Location
Chr.X: 8123301 - 8132858 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290783.1 NP_001277712.1 6 - 7 1507 66
NM_026137.5 NP_080413.2 6 - 7 1541 66
NM_028542.1 NP_082818.1 6 - 7 1793 66
GenBank mRNA AF220146.1 - 5 - 6 1063 66
AF353243.1 - 6 - 7 1187 66
AF513713.1 - 6 - 7 1010 66
AK006837.1 - 5 - 6 1029 66
AK017549.1 - 6 - 7 1123 66
BC057366.1 - 6 - 7 1507 66
BC080853.1 - 6 - 7 1456 66
BC108332.1 - 6 - 7 1297 66
BC120553.1 - 6 - 7 1044 66
BC125287.1 - 6 - 7 1043 66

View Full Product Details