Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr13
Entrez Gene ID
73447
Gene Name
WD repeat domain 13
Gene Aliases
1700060B08Rik, 5730411P10Rik, DXHXS7467e, W51679, mMg21
Chromosome Location
Chr.X: 8123301 - 8132858 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290783.1 NP_001277712.1 8 - 9 1773 97
NM_026137.5 NP_080413.2 8 - 9 1807 97
NM_028542.1 NP_082818.1 8 - 9 2059 97
GenBank mRNA AF220146.1 - 7 - 8 1329 97
AF353243.1 - 8 - 9 1453 97
AF513713.1 - 8 - 9 1276 97
AK006837.1 - 7 - 8 1295 97
AK017549.1 - 8 - 9 1389 97
BC080853.1 - 8 - 9 1722 97
BC108332.1 - 8 - 9 1563 97
BC120553.1 - 8 - 9 1310 97
BC125287.1 - 8 - 9 1309 97

View Full Product Details