Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dbn1
Entrez Gene ID
56320
Gene Name
drebrin 1
Gene Aliases
-
Chromosome Location
Chr.13: 55473428 - 55488163 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177371.1 NP_001170842.1 12 - 13 1399 112
NM_001177372.1 NP_001170843.1 11 - 12 1261 112
NM_019813.4 NP_062787.2 11 - 12 1261 112
GenBank mRNA AB064321.1 - 11 - 12 1708 112
AF187147.1 - 12 - 13 1389 112
AF187148.1 - 11 - 12 1251 112
AK132060.1 - 11 - 12 1247 112
AK162697.1 - 11 - 12 1248 112
BC006714.1 - 12 - 13 1209 112

View Full Product Details