The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032465.2 | NP_115854.2 | 14 - 15 | 1858 | 77 |
GenBank mRNA | AK007152.1 | - | 2 - 3 | 174 | 77 |
AK040539.1 | - | 14 - 15 | 1862 | 77 | |
AK156470.1 | - | 14 - 15 | 1859 | 77 | |
AY029156.1 | - | 14 - 15 | 1716 | 77 | |
BC052865.1 | - | 14 - 15 | 1755 | 77 |