The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017429.2 | NP_001017429.1 | 1 - 2 | 212 | 66 |
GenBank mRNA | AK146388.1 | - | 1 - 2 | 169 | 66 |
AK148493.1 | - | 1 - 2 | 180 | 66 | |
AK152968.1 | - | 1 - 2 | 173 | 66 | |
AK158051.1 | - | 1 - 2 | 190 | 66 | |
BC048668.1 | - | 1 - 2 | 212 | 66 | |
BG792513.1 | - | 1 - 2 | 144 | 66 |