The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001045521.1 | NP_001038986.1 | 4 - 5 | 736 | 81 |
GenBank mRNA | AK145810.1 | - | 4 - 5 | 741 | 81 |
BC147400.1 | - | 4 - 5 | 732 | 81 | |
BC147401.1 | - | 4 - 5 | 732 | 81 | |
BY757078.1 | - | 2 - 3 | 192 | 81 |