The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172493.2 | NP_766081.1 | 26 - 27 | 3478 | 67 |
GenBank mRNA | AB713187.1 | - | 27 - 28 | 3273 | 67 |
AK083254.1 | - | 19 - 20 | 2410 | 67 | |
AY312280.1 | - | 26 - 27 | 3415 | 67 | |
BB430043.2 | - | 3 - 4 | 360 | 67 | |
Y15910.1 | - | 7 - 8 | 985 | 67 |