The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163160.1 | NP_001156632.1 | 1 - 2 | 209 | 67 |
GenBank mRNA | AK076830.1 | - | 1 - 2 | 210 | 67 |