Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Golm1
Entrez Gene ID
105348
Gene Name
golgi membrane protein 1
Gene Aliases
2310001L02Rik, AW125446, D030064E01Rik, GP73, Golph2, PSEC0257
Chromosome Location
Chr.13: 59634996 - 59675784 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035122.2 NP_001030294.1 1 - 2 60 69
GenBank mRNA AK133865.1 - 1 - 2 62 69
CK391733.1 - 1 - 2 60 69

View Full Product Details