Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adam26b
Entrez Gene ID
382007
Gene Name
a disintegrin and metallopeptidase domain 26B
Gene Aliases
-
Chromosome Location
Chr.8: 43519864 - 43528137 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009547.2 NP_001009547.1 1 - 2 150 142
GenBank mRNA AK015931.1 - 1 - 2 150 141

View Full Product Details