Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ceacam1
Entrez Gene ID
26365
Gene Name
carcinoembryonic antigen-related cell adhesion molecule 1
Gene Aliases
Bgp, Bgp1, C-CAM, CD66a, Cc1, Cea-1, Cea-7, Cea1, Cea7, Hv-2, Hv2, MHVR, MHVR1, Mhv-1, bb-1, mCEA1, mmCGM1, mmCGM1a, mmCGM2
Chromosome Location
Chr.7: 25461702 - 25477625 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039185.1 NP_001034274.1 5 - 6 1332 127
NM_001039186.1 NP_001034275.1 5 - 6 1332 127
NM_001039187.1 NP_001034276.1 3 - 4 792 127
NM_011926.2 NP_036056.2 3 - 4 792 127
GenBank mRNA AB236328.2 - 3 - 4 701 127
AB236329.1 - 3 - 4 701 127
AB236330.2 - 4 - 5 986 127
BC016891.1 - 5 - 6 1284 127
BC114998.1 - 1 - 2 264 127
BC121189.1 - 4 - 5 889 127
M77196.1 - 5 - 6 1310 127
M96934.1 - 5 - 6 1258 127
M96935.1 - 4 - 5 694 127
X15351.1 - 5 - 6 1344 127
X53084.1 - 3 - 4 721 127
X67278.1 - 3 - 4 804 127
X67279.1 - 5 - 6 1344 127
X67280.1 - 5 - 6 1261 127
X67281.1 - 5 - 6 1261 127
X67282.1 - 3 - 4 804 127
X67283.1 - 3 - 4 721 127

View Full Product Details