The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021330.4 | NP_067305.2 | 3 - 4 | 307 | 122 |
GenBank mRNA | AK082955.1 | - | 3 - 4 | 252 | 122 |
AK167887.1 | - | 3 - 4 | 267 | 122 | |
BC096368.1 | - | 3 - 4 | 251 | 122 | |
BC111893.1 | - | 3 - 4 | 246 | 122 | |
BC132258.1 | - | 3 - 4 | 246 | 122 | |
Y17345.1 | - | 3 - 4 | 227 | 122 |