The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008138.4 | NP_032164.2 | 1 - 1 | 48 | 134 |
GenBank mRNA | AK157998.1 | - | 1 - 1 | 52 | 134 |
AK159222.1 | - | 1 - 1 | 52 | 134 | |
AK167388.1 | - | 1 - 1 | 50 | 134 |