Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Nacc2
- Entrez Gene ID
-
67991
- Gene Name
- nucleus accumbens associated 2, BEN and BTB (POZ) domain containing
- Gene Aliases
-
0610020I02Rik, AI448087, Btbd14a, C030048H19Rik
- Chromosome Location
-
Chr.2: 26055535 - 26122811 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK076376.1 |
- |
- |
|
151 |
View Full Product Details