Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY560673.1 - 1 346 Chr. 12 - 74035344 - 74035727 [-] on Build GRCm38 76
GenBankmRNA BY566653.1 - 1 431 Chr. 12 - 74035344 - 74035801 [-] on Build GRCm38 76
GenBankmRNA AK157120.1 - 1122 Chr. - - [-] on Build 76
GenBankmRNA BY572042.1 - 1 413 Chr. 12 - 74035344 - 74035783 [-] on Build GRCm38 76
GenBankmRNA BY559710.1 - 1 353 Chr. 12 - 74035344 - 74035734 [-] on Build GRCm38 76
GenBankmRNA BY564748.1 - 1 415 Chr. 12 - 74035344 - 74035797 [-] on Build GRCm38 76
GenBankmRNA BY765124.1 - 2 687 Chr. 12 - 74035344 - 74036375 [-] on Build GRCm38 76


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream synaptotagmin XVI Syt16 6430703N11, Strep14, Syt14l, syt14r Anti-sense Chr. - 73997608 - 74267916 on Build GRCm38
Downstream RIKEN cDNA 1700086L19 gene 1700086L19Rik 1700006L05Rik, AU016007 248.5kb Anti-sense Chr. - 74284276 - 74295950 on Build GRCm38

View Full Product Details