The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205385.1 | NP_001192314.1 | 7 - 8 | 749 | 96 |
NM_001205386.1 | NP_001192315.1 | 7 - 8 | 746 | 96 | |
NM_023735.2 | NP_076224.1 | 6 - 7 | 941 | 96 | |
GenBank mRNA | AK004554.1 | - | 6 - 7 | 755 | 96 |
AK083343.1 | - | 6 - 7 | 759 | 96 | |
AK145536.1 | - | 6 - 7 | 761 | 96 | |
AK150487.1 | - | 6 - 7 | 643 | 96 | |
AK150589.1 | - | 6 - 7 | 719 | 96 | |
AK150991.1 | - | 6 - 7 | 699 | 96 | |
AK152865.1 | - | 6 - 7 | 736 | 96 | |
AK153079.1 | - | 6 - 7 | 762 | 96 | |
AK153150.1 | - | 6 - 7 | 736 | 96 | |
AK153204.1 | - | 6 - 7 | 642 | 96 | |
AK159135.1 | - | 6 - 7 | 728 | 96 | |
AK159668.1 | - | 6 - 7 | 653 | 96 | |
AK159775.1 | - | 6 - 7 | 740 | 96 | |
AK160047.1 | - | 6 - 7 | 601 | 96 | |
AK166529.1 | - | 7 - 8 | 693 | 96 | |
AK166586.1 | - | 7 - 8 | 693 | 96 | |
AK167148.1 | - | 6 - 7 | 700 | 96 | |
AK167891.1 | - | 6 - 7 | 698 | 96 | |
AK168771.1 | - | 6 - 7 | 714 | 96 | |
AK187649.1 | - | 1 - 2 | 62 | 96 | |
BC005557.1 | - | 6 - 7 | 704 | 96 | |
BC080806.1 | - | 7 - 8 | 707 | 96 | |
BC108386.1 | - | 6 - 7 | 672 | 96 |