The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025586.3 | NP_079862.1 | 4 - 4 | 556 | 163 |
GenBank mRNA | AK002234.1 | - | 4 - 4 | 553 | 163 |
AK010319.1 | - | 4 - 4 | 557 | 163 | |
AK010674.1 | - | 4 - 4 | 555 | 163 | |
AK010916.1 | - | 4 - 4 | 557 | 163 | |
AK010931.1 | - | 4 - 4 | 557 | 163 | |
AK012025.1 | - | 4 - 4 | 553 | 163 | |
AK012423.1 | - | 4 - 4 | 548 | 163 | |
AK019386.1 | - | 3 - 3 | 544 | 163 | |
AK151138.1 | - | 4 - 4 | 553 | 163 | |
AK152711.1 | - | 4 - 4 | 554 | 163 | |
AK159183.1 | - | 4 - 4 | 554 | 163 | |
AK159626.1 | - | 4 - 4 | 554 | 163 | |
AK167996.1 | - | 4 - 4 | 554 | 163 | |
AK168125.1 | - | 4 - 4 | 557 | 163 | |
AK168807.1 | - | 3 - 3 | 548 | 163 | |
AK169662.1 | - | 4 - 4 | 554 | 163 | |
AK186686.1 | - | 2 - 2 | 320 | 163 | |
BC079842.1 | - | 4 - 4 | 543 | 163 | |
BC081441.1 | - | 3 - 3 | 540 | 163 | |
BC081442.1 | - | 3 - 3 | 536 | 163 | |
BC087917.1 | - | 4 - 4 | 583 | 163 | |
BC091735.1 | - | 3 - 3 | 533 | 163 | |
BC100335.1 | - | 3 - 3 | 523 | 163 | |
BC100561.1 | - | - | 828 | 163 |