The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011507.3 | NP_035637.2 | 1 - 2 | 161 | 101 |
GenBank mRNA | AK042987.1 | - | 1 - 2 | 116 | 101 |
AK045176.1 | - | 1 - 2 | 114 | 101 | |
AK156263.1 | - | 1 - 2 | 116 | 101 | |
AK167136.1 | - | 1 - 2 | 116 | 101 | |
AK172363.1 | - | 1 - 2 | 116 | 101 | |
AK172561.1 | - | 1 - 2 | 116 | 101 | |
AK172633.1 | - | 1 - 2 | 116 | 101 | |
BC080781.1 | - | 1 - 2 | 116 | 101 | |
BI985474.1 | - | 1 - 2 | 153 | 102 |