The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009098.2 | NP_033124.1 | 6 - 6 | 683 | 86 |
GenBank mRNA | AK009023.1 | - | 6 - 6 | 636 | 86 |
AK010650.1 | - | 6 - 6 | 683 | 86 | |
AK011048.1 | - | 6 - 6 | 652 | 86 | |
AK011058.1 | - | 6 - 6 | 636 | 86 | |
AK011642.1 | - | 6 - 6 | 628 | 86 | |
AK012435.1 | - | 6 - 6 | 636 | 86 | |
AK088660.1 | - | 6 - 6 | 636 | 86 | |
AK146806.1 | - | 6 - 6 | 636 | 86 | |
AK166848.1 | - | 6 - 6 | 636 | 86 | |
AK168609.1 | - | 6 - 6 | 636 | 86 | |
BC027217.1 | - | 5 - 5 | 615 | 86 | |
BC051446.1 | - | 5 - 5 | 632 | 86 | |
BC081465.1 | - | 5 - 5 | 615 | 86 | |
BC086899.1 | - | 6 - 6 | 635 | 86 | |
BC100594.1 | - | 6 - 6 | 635 | 86 | |
X73829.1 | - | 5 - 5 | 618 | 87 |