The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166427.1 | NP_001159899.1 | 3 - 3 | 2053 | 147 |
NM_001166428.1 | NP_001159900.1 | 4 - 4 | 1992 | 147 | |
NM_001166429.1 | NP_001159901.1 | 4 - 4 | 1953 | 147 | |
NM_001166430.1 | NP_001159902.1 | 4 - 4 | 2304 | 147 | |
NM_001166431.1 | NP_001159903.1 | 3 - 3 | 2243 | 147 | |
NM_001166432.1 | NP_001159904.1 | 4 - 4 | 2044 | 147 | |
NM_133834.2 | NP_598595.1 | 4 - 4 | 1935 | 147 | |
GenBank mRNA | AK076478.1 | - | 3 - 3 | 1893 | 147 |
AK190486.1 | - | 1 - 1 | 153 | 147 | |
BC018185.1 | - | 3 - 3 | 1878 | 148 | |
BC023162.1 | - | 1 - 1 | 959 | 148 | |
BC023793.1 | - | 4 - 4 | 1930 | 148 | |
BC025465.1 | - | 1 - 1 | 669 | 148 | |
BC025481.1 | - | 4 - 4 | 1912 | 148 | |
BC027003.1 | - | 1 - 1 | 1512 | 147 | |
BC029163.1 | - | 1 - 1 | 1696 | 148 | |
BC029764.1 | - | 1 - 1 | 1519 | 148 | |
BC032197.1 | - | 4 - 4 | 1930 | 148 | |
BC033483.1 | - | 1 - 1 | 1694 | 148 | |
BC042689.1 | - | 3 - 3 | 2176 | 147 | |
BC047346.1 | - | 3 - 3 | 2176 | 147 | |
BC089313.1 | - | 4 - 4 | 1777 | 149 | |
CX218091.1 | - | 1 - 1 | 172 | 147 |