The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198193.2 | NP_937836.1 | 4 - 4 | 360 | 115 |
GenBank mRNA | AK012289.1 | - | 4 - 4 | 360 | 115 |
AK131854.1 | - | 6 - 6 | 663 | 115 | |
AY054973.1 | - | 2 - 2 | 226 | 115 | |
AY056835.1 | - | 2 - 2 | 226 | 115 | |
BC132308.1 | - | 2 - 2 | 263 | 115 | |
FJ594067.1 | - | 5 - 5 | 464 | 115 | |
FJ594068.1 | - | 4 - 4 | 394 | 115 |