Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phkg1
Entrez Gene ID
18682
Gene Name
phosphorylase kinase gamma 1
Gene Aliases
Phkg
Chromosome Location
Chr.5: 129863435 - 129879083 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011079.2 NP_035209.1 5 - 6 569 157
GenBank mRNA BC055102.1 - 5 - 6 569 157
J03293.1 - 5 - 6 439 157
M16216.1 - 4 - 5 407 157

View Full Product Details