The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033979.1 | NP_001029151.1 | 2 - 3 | 251 | 87 |
GenBank mRNA | AK132881.1 | - | 2 - 3 | 251 | 87 |