The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009076.3 | NP_033102.2 | 2 - 3 | 199 | 62 |
GenBank mRNA | AK002973.1 | - | 2 - 3 | 198 | 62 |
AK008347.1 | - | 2 - 3 | 198 | 62 | |
AK012349.1 | - | 2 - 3 | 198 | 62 | |
AK012428.1 | - | 2 - 3 | 196 | 62 | |
AK088464.1 | - | 2 - 3 | 198 | 62 | |
AK144905.1 | - | 2 - 3 | 198 | 62 | |
AK148566.1 | - | - | 400 | 62 | |
AK150318.1 | - | 2 - 3 | 199 | 62 | |
AK151532.1 | - | 2 - 3 | 198 | 62 | |
AK151592.1 | - | 2 - 3 | 198 | 62 | |
AK167761.1 | - | 2 - 3 | 218 | 62 | |
AK168121.1 | - | 2 - 3 | 198 | 62 | |
BC018321.1 | - | 2 - 3 | 195 | 62 | |
BC075731.1 | - | 1 - 2 | 387 | 62 | |
BC081469.1 | - | 2 - 3 | 182 | 62 | |
BC090393.1 | - | - | 177 | 62 | |
BY295120.1 | - | 2 - 3 | 62 | ||
L04280.1 | - | 2 - 3 | 62 |