Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Defb47
Entrez Gene ID
654465
Gene Name
defensin beta 47
Gene Aliases
Defb41, EG654465
Chromosome Location
Chr.14: 62992375 - 63001180 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039125.2 NP_001034214.1 1 - 2 78 69
GenBank mRNA BC132604.1 - 1 - 2 78 69
BC132606.1 - 1 - 2 78 69
DQ012043.1 - 1 - 2 53 69

View Full Product Details