The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077529.2 | NP_001070997.1 | 1 - 2 | 288 | 96 |
NM_008705.5 | NP_032731.1 | 2 - 3 | 560 | 96 | |
GenBank mRNA | AK012447.1 | - | 2 - 3 | 217 | 96 |
AK150419.1 | - | 2 - 3 | 222 | 96 | |
AK151262.1 | - | 1 - 2 | 177 | 96 | |
AK169120.1 | - | 2 - 3 | 182 | 96 | |
AV458476.1 | - | 1 - 2 | 288 | 96 | |
BC066995.1 | - | 2 - 3 | 560 | 96 | |
BC086892.1 | - | 1 - 2 | 193 | 96 | |
BC086893.1 | - | 2 - 3 | 185 | 96 | |
BY274368.1 | - | 2 - 3 | 311 | 96 | |
EF437324.1 | - | 1 - 2 | 108 | 96 | |
X68193.1 | - | 1 - 2 | 144 | 96 |