The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018871.3 | NP_061359.2 | 1 - 2 | 311 | 69 |
GenBank mRNA | AB073970.1 | - | 1 - 2 | 258 | 69 |
AK088847.1 | - | 1 - 2 | 224 | 69 | |
AK148618.1 | - | 1 - 2 | 270 | 69 | |
AK153307.1 | - | 1 - 2 | 270 | 69 | |
AK164356.1 | - | 1 - 2 | 262 | 69 | |
AK183704.1 | - | 1 - 2 | 47 | 69 | |
BC008129.1 | - | 1 - 2 | 194 | 69 | |
BY244024.1 | - | 1 - 2 | 326 | 69 | |
CT010208.1 | - | 1 - 2 | 79 | 69 |