Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn1r75
Entrez Gene ID
171241
Gene Name
vomeronasal 1 receptor 75
Gene Aliases
V1rg6
Chromosome Location
Chr.7: 11880343 - 11881260 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134207.1 NP_598968.1 1 - 1 871 80
GenBank mRNA BC115506.1 - 1 - 1 864 80
BC115507.1 - 1 - 1 871 80

View Full Product Details