The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025364.2 | NP_079640.1 | 10 - 11 | 754 | 95 |
GenBank mRNA | AK003453.1 | - | 10 - 11 | 606 | 95 |
AK088266.1 | - | 10 - 11 | 603 | 95 | |
AK146248.1 | - | 10 - 11 | 632 | 95 | |
BC027510.1 | - | 10 - 11 | 593 | 95 | |
BC094393.1 | - | 11 - 12 | 785 | 95 | |
BC157909.1 | - | 10 - 11 | 618 | 95 | |
BC158023.1 | - | 10 - 11 | 618 | 95 | |
BC172063.1 | - | 10 - 11 | 615 | 95 | |
BF148013.1 | - | 1 - 2 | 95 |