Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gli3
Entrez Gene ID
14634
Gene Name
GLI-Kruppel family member GLI3
Gene Aliases
AI854843, AU023367, Bph, GLI3-190, GLI3FL, Pdn, Xt, add
Chromosome Location
Chr.13: 15463235 - 15730026 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008130.2 NP_032156.2 15 - 15 8331 124
GenBank mRNA AK048009.1 - 1 - 1 2780 124

View Full Product Details