Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mup7
Entrez Gene ID
100041658
Gene Name
major urinary protein 7
Gene Aliases
Gm12546, Mup3, OTTMUSG00000007428
Chromosome Location
Chr.4: 60066469 - 60070481 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134675.1 NP_001128147.1 1 - 1 42 101

View Full Product Details